siRNA Details
| Gene Symbol: Rhog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037195.1 | NP_001032272.1 | 2 | 168 | ||
| XM_006229882.2 | XP_006229944.1 | 3 | 309 | |||
| XM_006229883.2 | XP_006229945.1 | 2 | 415 | |||
| GenBank mRNA | BC107943.1 | - | 2 | 168 | 7389075 | |
| FQ212590.1 | - | 2 | 208 | |||
| FQ232547.1 | - | 2 | 211 | |||

Rhog
Genomic Map