siRNA Details
| Gene Symbol: Mrvi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105210.1 | NP_001098680.1 | 19 | 2709 | ||
| NM_001105211.1 | NP_001098681.1 | 18 | 2462 | |||
| XM_006230047.2 | XP_006230109.1 | 19 | 2707 | |||
| XM_006230048.2 | XP_006230110.1 | 20 | 2893 | |||
| XM_008759727.1 | XP_008757949.1 | 19 | 2809 | |||
| XM_008759728.1 | XP_008757950.1 | 19 | 3028 | |||
| XM_008759729.1 | XP_008757951.1 | 20 | 2490 | |||

Mrvi1
Genomic Map