siRNA Details
| Gene Symbol: Ubqln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107543.2 | NP_001101013.1 | 2 | 1081 | ||
| GenBank mRNA | BC161885.1 | - | 2 | 1081 | 7133921 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubqln3| Gene Symbol: Ubqln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107543.2 | NP_001101013.1 | 2 | 1081 | ||
| GenBank mRNA | BC161885.1 | - | 2 | 1081 | 7133921 | |