siRNA Details
| Gene Symbol: Rrp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008346.1 | NP_001008347.1 | 3 | 546 | ||
| GenBank mRNA | BC085119.1 | - | 3 | 546 | 7108537 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rrp8| Gene Symbol: Rrp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008346.1 | NP_001008347.1 | 3 | 546 | ||
| GenBank mRNA | BC085119.1 | - | 3 | 546 | 7108537 | |