siRNA Details
| Gene Symbol: Dchs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107544.2 | NP_001101014.2 | 5 | 3323 | ||
| XM_006229942.2 | XP_006230004.1 | 6 | 3512 | |||
| XM_008759732.1 | XP_008757954.1 | 6 | 6234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dchs1| Gene Symbol: Dchs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107544.2 | NP_001101014.2 | 5 | 3323 | ||
| XM_006229942.2 | XP_006230004.1 | 6 | 3512 | |||
| XM_008759732.1 | XP_008757954.1 | 6 | 6234 | |||