siRNA Details
| Gene Symbol: Pdzd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024777.2 | NP_001019948.2 | 5 | 625 | ||
| XM_008759741.1 | XP_008757963.1 | 4 | 2860 | |||
| XM_008759742.1 | XP_008757964.1 | 5 | 830 | |||
| XM_008759743.1 | XP_008757965.1 | 4 | 536 | |||
| XM_008759744.1 | XP_008757966.1 | 6 | 3084 | |||
| GenBank mRNA | AY387091.1 | - | 5 | 625 | ||
| BC097426.1 | - | 4 | 522 | 7455141 | ||

Pdzd9
Genomic Map