siRNA Details
| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230198.2 | XP_006230260.1 | 17 | 5850 | ||
| XM_219296.6 | XP_219296.3 | 18 | 5953 | |||
| GenBank mRNA | BC089931.1 | - | 1 | 226 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp6| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230198.2 | XP_006230260.1 | 17 | 5850 | ||
| XM_219296.6 | XP_219296.3 | 18 | 5953 | |||
| GenBank mRNA | BC089931.1 | - | 1 | 226 | ||