siRNA Details
| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230198.2 | XP_006230260.1 | 8 | 1832 | ||
| XM_219296.6 | XP_219296.3 | 8 | 1833 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp6| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230198.2 | XP_006230260.1 | 8 | 1832 | ||
| XM_219296.6 | XP_219296.3 | 8 | 1833 | |||