siRNA Details
| Gene Symbol: Il21r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012469.1 | NP_001012487.1 | 10 | 1437 | ||
| XM_006230261.2 | XP_006230323.1 | 9 | 1474 | |||
| XM_006230262.2 | XP_006230324.1 | 10 | 1450 | |||
| GenBank mRNA | BC089857.1 | - | 10 | 1437 | 7381492 | |
| FQ226709.1 | - | 9 | 1308 | |||

Il21r
Genomic Map