siRNA Details
| Gene Symbol: Tmem219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748988.3 | XP_003749036.1 | 6, 7 | 1252 | ||
| XM_008759928.1 | XP_008758150.1 | 5, 6 | 938 | |||
| XM_008759929.1 | XP_008758151.1 | 4, 5 | 743 | |||
| XM_008759930.1 | XP_008758152.1 | 4, 5 | 740 | |||
| GenBank mRNA | FQ216353.1 | - | 2, 3 | 650 | ||

Tmem219
Genomic Map