siRNA Details
| Gene Symbol: LOC308990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025001.1 | NP_001020172.1 | 2 | 476 | ||
| XM_006230265.2 | XP_006230327.1 | 2 | 279 | |||
| XM_006230266.2 | XP_006230328.1 | 2 | 420 | |||
| XM_006230267.2 | XP_006230329.1 | 2 | 378 | |||
| XM_006230268.2 | XP_006230330.1 | 2 | 237 | |||
| GenBank mRNA | BC091205.1 | - | 1 | 259 | 7380611 | |
| BC091216.1 | - | 2 | 476 | 7383904 | ||

LOC308990
Genomic Map