siRNA Details
| Gene Symbol: Sephs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079889.2 | NP_001073358.2 | 1 | 1441 | ||
| GenBank mRNA | BC081915.1 | - | 1 | 1336 | ||
| BC097429.1 | - | 1 | 1441 | |||
| BC128783.1 | - | 1 | 1298 | 7378558 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sephs2| Gene Symbol: Sephs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079889.2 | NP_001073358.2 | 1 | 1441 | ||
| GenBank mRNA | BC081915.1 | - | 1 | 1336 | ||
| BC097429.1 | - | 1 | 1441 | |||
| BC128783.1 | - | 1 | 1298 | 7378558 | ||