siRNA Details
| Gene Symbol: Zfp629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107551.2 | NP_001101021.2 | 3 | 2482 | ||
| XM_006230272.2 | XP_006230334.1 | 3 | 2411 | |||
| GenBank mRNA | AY589489.1 | - | 3 | 2472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp629| Gene Symbol: Zfp629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107551.2 | NP_001101021.2 | 3 | 2482 | ||
| XM_006230272.2 | XP_006230334.1 | 3 | 2411 | |||
| GenBank mRNA | AY589489.1 | - | 3 | 2472 | ||