siRNA Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230276.2 | XP_006230338.1 | 5 | 910 | ||
| XM_006230277.2 | XP_006230339.1 | 5 | 910 | |||
| XM_006230280.2 | XP_006230342.1 | 5 | 794 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl19| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230276.2 | XP_006230338.1 | 5 | 910 | ||
| XM_006230277.2 | XP_006230339.1 | 5 | 910 | |||
| XM_006230280.2 | XP_006230342.1 | 5 | 794 | |||