siRNA Details
| Gene Symbol: Fbxl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107552.1 | NP_001101022.1 | 3 | 454 | ||
| XM_006230276.2 | XP_006230338.1 | 4 | 802 | |||
| XM_006230277.2 | XP_006230339.1 | 4 | 802 | |||
| XM_006230278.2 | XP_006230340.1 | 4 | 802 | |||
| XM_006230279.2 | XP_006230341.1 | 4 | 802 | |||
| XM_006230280.2 | XP_006230342.1 | 4 | 686 | |||

Fbxl19
Genomic Map