siRNA Details
| Gene Symbol: Vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203335.2 | NP_976080.1 | 2 | 258 | ||
| GenBank mRNA | AY423047.1 | - | 2 | 241 | ||
| BC166413.1 | - | 254 | 9100951 | |||
| CB314647.1 | - | 2 | 274 | |||
| FQ210919.1 | - | 2 | 300 | |||

Vkorc1
Genomic Map