siRNA Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134859.1 | NP_001128331.1 | 13 | 2423 | ||
| XM_006230387.1 | XP_006230449.1 | 13 | 2357 | |||
| XM_006230388.1 | XP_006230450.1 | 12 | 1924 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec23ip| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134859.1 | NP_001128331.1 | 13 | 2423 | ||
| XM_006230387.1 | XP_006230449.1 | 13 | 2357 | |||
| XM_006230388.1 | XP_006230450.1 | 12 | 1924 | |||