siRNA Details
| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230424.2 | XP_006230486.1 | 24 | 3281 | ||
| XM_219377.8 | XP_219377.5 | 24 | 3280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr11| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230424.2 | XP_006230486.1 | 24 | 3281 | ||
| XM_219377.8 | XP_219377.5 | 24 | 3280 | |||