siRNA Details
Gene Symbol: Wdr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230424.2 | XP_006230486.1 | 22, 23 | 3064 | ||
XM_219377.8 | XP_219377.5 | 22, 23 | 3063 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230424.2 | XP_006230486.1 | 22, 23 | 3064 | ||
XM_219377.8 | XP_219377.5 | 22, 23 | 3063 |