siRNA Details
| Gene Symbol: Tacc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004415.2 | NP_001004415.1 | 7608 | |||
| NM_001004418.2 | NP_001004418.1 | 2493 | ||||
| XM_006230398.2 | XP_006230460.1 | 7, 8 | 2589 | |||
| XM_008759883.1 | XP_008758105.1 | 7, 8 | 2589 | |||
| XM_008759884.1 | XP_008758106.1 | 8471 | ||||
| GenBank mRNA | FQ223932.1 | - | 5, 6 | 963 | ||
| KJ143743.1 | - | 7, 8 | 1959 | |||

Tacc2
Genomic Map