siRNA Details
| Gene Symbol: RGD1305014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014025.1 | NP_001014047.1 | 2, 3 | 370 | ||
| XM_008759885.1 | XP_008758107.1 | 2, 3 | 1572 | |||
| GenBank mRNA | BC083846.1 | - | 2, 3 | 370 | 7121351 | |

RGD1305014
Genomic Map