siRNA Details
| Gene Symbol: Ikzf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107555.1 | NP_001101025.1 | 5 | 952 | ||
| XM_006230401.1 | XP_006230463.1 | 4 | 789 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikzf5| Gene Symbol: Ikzf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107555.1 | NP_001101025.1 | 5 | 952 | ||
| XM_006230401.1 | XP_006230463.1 | 4 | 789 | |||