siRNA Details
| Gene Symbol: RGD1307235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134546.1 | NP_001128018.1 | 24 | 3336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307235| Gene Symbol: RGD1307235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134546.1 | NP_001128018.1 | 24 | 3336 | ||