siRNA Details
| Gene Symbol: Adam12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759968.1 | XP_008758190.1 | 22 | 2817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam12| Gene Symbol: Adam12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759968.1 | XP_008758190.1 | 22 | 2817 | ||