siRNA Details
| Gene Symbol: Clrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014026.1 | NP_001014048.1 | 346 | |||
| GenBank mRNA | BC078942.1 | - | 346 | 7109328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clrn3| Gene Symbol: Clrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014026.1 | NP_001014048.1 | 346 | |||
| GenBank mRNA | BC078942.1 | - | 346 | 7109328 | ||