siRNA Details
Gene Symbol: Gpr123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760017.1 | XP_008758239.1 | 7 | 2667 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
