siRNA Details
| Gene Symbol: Scgb1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107561.1 | NP_001101031.1 | 3 | 420 | ||
| GenBank mRNA | X60661.1 | - | 3 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1c1| Gene Symbol: Scgb1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107561.1 | NP_001101031.1 | 3 | 420 | ||
| GenBank mRNA | X60661.1 | - | 3 | 399 | ||