siRNA Details
| Gene Symbol: Ifitm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760024.1 | XP_008758246.1 | 3 | 1518 | ||
| XM_008760025.1 | XP_008758247.1 | 3 | 220 | |||
| XM_219476.7 | XP_219476.5 | 2 | 1721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm6| Gene Symbol: Ifitm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760024.1 | XP_008758246.1 | 3 | 1518 | ||
| XM_008760025.1 | XP_008758247.1 | 3 | 220 | |||
| XM_219476.7 | XP_219476.5 | 2 | 1721 | |||