siRNA Details
| Gene Symbol: Tnpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100692.1 | NP_001094162.1 | 17, 18 | 2126 | ||
| XM_006231839.2 | XP_006231901.1 | 18, 19 | 2717 | |||
| XM_006231840.2 | XP_006231902.1 | 18, 19 | 2441 | |||
| XM_008760668.1 | XP_008758890.1 | 18, 19 | 2729 | |||
| GenBank mRNA | BC090323.1 | - | 15, 16 | 1806 | ||

Tnpo1
Genomic Map