siRNA Details
| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107566.1 | NP_001101036.1 | 7 | 735 | ||
| XM_006230784.2 | XP_006230846.1 | 7 | 758 | |||
| GenBank mRNA | BC166782.1 | - | 7 | 671 | 7935012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpcn2| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107566.1 | NP_001101036.1 | 7 | 735 | ||
| XM_006230784.2 | XP_006230846.1 | 7 | 758 | |||
| GenBank mRNA | BC166782.1 | - | 7 | 671 | 7935012 | |