siRNA Details
| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107566.1 | NP_001101036.1 | 21, 22 | 1968 | ||
| XM_006230784.2 | XP_006230846.1 | 22, 23 | 2057 | |||
| GenBank mRNA | BC166782.1 | - | 22, 23 | 2032 | 7935012 | |

Tpcn2
Genomic Map