siRNA Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012069.2 | NP_001012069.2 | 6, 7 | 967 | ||
| GenBank mRNA | BC083920.1 | - | 6, 7 | 953 | 7135090 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtl5| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012069.2 | NP_001012069.2 | 6, 7 | 967 | ||
| GenBank mRNA | BC083920.1 | - | 6, 7 | 953 | 7135090 | |