siRNA Details
| Gene Symbol: RGD1311946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025683.1 | NP_001020854.1 | 4 | 1710 | ||
| GenBank mRNA | BC098053.1 | - | 4 | 1710 | 7457583 | |
| FQ229270.1 | - | 1 | 681 | |||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Mouse: NM_028077, AK007868, AK154828, BC019471.
					            	This siRNA also targets the following transcripts in Mouse: NM_028077, AK007868, AK154828, BC019471. Back To Top
 Back To Top
 RGD1311946
RGD1311946 Genomic Map
Genomic Map Back To Top
 Back To Top