siRNA Details
| Gene Symbol: Tvp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107568.3 | NP_001101038.2 | 4 | 312 | ||
| GenBank mRNA | BC168178.1 | - | 4 | 310 | ||
| CK483802.1 | - | 4 | 312 | |||
| FQ212667.1 | - | 4 | 348 | |||
| FQ219631.1 | - | 4 | 313 | |||

Tvp23b
Genomic Map