siRNA Details
| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199267.2 | NP_954888.1 | 11 | 1551 | ||
| GenBank mRNA | AY307375.1 | - | 10 | 1490 | ||
| BC079457.1 | - | 11 | 1551 | 7121610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rela| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199267.2 | NP_954888.1 | 11 | 1551 | ||
| GenBank mRNA | AY307375.1 | - | 10 | 1490 | ||
| BC079457.1 | - | 11 | 1551 | 7121610 | ||