siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230947.2 | XP_006231009.1 | 26 | 4619 | ||
| XM_006230948.2 | XP_006231010.1 | 26 | 4616 | |||
| XM_006230950.2 | XP_006231012.1 | 26 | 4589 | |||
| XM_006230952.2 | XP_006231014.1 | 26 | 4550 | |||
| XM_219515.8 | XP_219515.4 | 26 | 4580 | |||

Pcnxl3
Genomic Map