siRNA Details
| Gene Symbol: Ppp2r5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181379.2 | NP_852044.1 | 12 | 1380 | ||
| XM_008760143.1 | XP_008758365.1 | 13 | 1951 | |||
| GenBank mRNA | AY251278.1 | - | 12 | 1333 | ||
| BC090324.1 | - | 12 | 1380 | 7128996 | ||

Ppp2r5b
Genomic Map