siRNA Details
| Gene Symbol: SPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007226.1 | NP_001007227.1 | 9, 10 | 1193 | ||
| NM_001007227.1 | NP_001007228.1 | 8, 9 | 1071 | |||
| NM_001007228.1 | NP_001007229.1 | 7, 8 | 1019 | |||
| NM_001007229.1 | NP_001007230.1 | 8, 9 | 1070 | |||
| NM_001007230.1 | NP_001007231.1 | 9, 10 | 1122 | |||
| NM_003563.3 | NP_003554.1 | 8, 9 | 1141 | |||
| XM_005257723.4 | XP_005257780.1 | 8, 9 | 1365 | |||
| XM_005257724.4 | XP_005257781.1 | 7, 8 | 1295 | |||
| XM_017025204.1 | XP_016880693.1 | 6, 7 | 808 | |||
| GenBank mRNA | AJ000644.1 | - | 7, 8 | 879 | ||
| AK293045.1 | - | 8, 9 | 1352 | |||
| AK293587.1 | - | 7, 8 | 825 | |||
| AK300616.1 | - | 7, 8 | 843 | |||
| AK303268.1 | - | 6, 7 | 765 | |||
| AK312691.1 | - | 7, 8 | 855 | |||
| AK316366.1 | - | 9, 10 | 920 | |||
| BC001269.1 | - | 8, 9 | 1025 | 3461565 | ||
| BC003385.1 | - | 7, 8 | 836 | 3445878 | ||

SPOP
Genomic Map