siRNA Details
| Gene Symbol: Dagla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005886.1 | NP_001005886.1 | 12, 13 | 1393 | ||
| XM_008760229.1 | XP_008758451.1 | 11, 12 | 4763 | |||
| XM_008760230.1 | XP_008758452.1 | 12, 13 | 4678 | |||
| GenBank mRNA | AY275375.1 | - | 12, 13 | 1393 | ||

Dagla
Genomic Map