siRNA Details
| Gene Symbol: Olr318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000505.1 | NP_001000505.1 | 1 | 626 | ||
| Gene Symbol: Olr1874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728868.4 | XP_002728914.2 | 1 | 1231 | ||

Olr318
Genomic Map