siRNA Details
Gene Symbol: Olr337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000507.1 | NP_001000507.1 | 1 | 496 | ||
GenBank mRNA | AF091562.1 | - | 1 | 304 | ||
AF091564.1 | - | 1 | 304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000507.1 | NP_001000507.1 | 1 | 496 | ||
GenBank mRNA | AF091562.1 | - | 1 | 304 | ||
AF091564.1 | - | 1 | 304 |