siRNA Details
| Gene Symbol: Otud7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759566.1 | XP_008757788.1 | 4 | 892 | ||
| XM_008759567.1 | XP_008757789.1 | 4 | 1173 | |||
| XM_008759568.1 | XP_008757790.1 | 4 | 892 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud7a| Gene Symbol: Otud7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759566.1 | XP_008757788.1 | 4 | 892 | ||
| XM_008759567.1 | XP_008757789.1 | 4 | 1173 | |||
| XM_008759568.1 | XP_008757790.1 | 4 | 892 | |||