siRNA Details
Gene Symbol: Trex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107580.1 | NP_001101050.1 | 2 | 160 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107580.1 | NP_001101050.1 | 2 | 160 |