siRNA Details
| Gene Symbol: Plxna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107581.2 | NP_001101051.2 | 19 | 3400 | ||
| XM_006229577.2 | XP_006229639.1 | 19 | 3559 | |||
| XM_006229578.2 | XP_006229640.1 | 19 | 3816 | |||
| XM_008773598.1 | XP_008771820.1 | 18 | 5308 | |||

Plxna3
Genomic Map