siRNA Details
| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107583.1 | NP_001101053.1 | 20 | 2668 | ||
| XM_006231210.2 | XP_006231272.1 | 24 | 3114 | |||
| GenBank mRNA | FQ209556.1 | - | 6 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gldc| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107583.1 | NP_001101053.1 | 20 | 2668 | ||
| XM_006231210.2 | XP_006231272.1 | 24 | 3114 | |||
| GenBank mRNA | FQ209556.1 | - | 6 | 626 | ||