siRNA Details
| Gene Symbol: Chuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107588.1 | NP_001101058.1 | 11 | 1279 | ||
| XM_006231481.2 | XP_006231543.1 | 11 | 1276 | |||
| XM_008760418.1 | XP_008758640.1 | 11 | 1277 | |||
| XM_008760419.1 | XP_008758641.1 | 11 | 1277 | |||
| GenBank mRNA | BC168905.1 | - | 11 | 1254 | 9088878 | |

Chuk
Genomic Map