siRNA Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199104.2 | NP_954535.2 | 8 | 1049 | ||
| XM_006231392.2 | XP_006231454.1 | 8 | 1065 | |||
| XM_008760422.1 | XP_008758644.1 | 7 | 1573 | |||
| XM_008760423.1 | XP_008758645.1 | 7 | 1573 | |||
| GenBank mRNA | BC061741.1 | - | 8 | 1015 | 5599023 | |

Zfyve27
Genomic Map