siRNA Details
| Gene Symbol: Pyroxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004261.1 | NP_001004261.1 | 8, 9 | 796 | ||
| XM_006231394.2 | XP_006231456.1 | 8, 9 | 1067 | |||
| XM_006231396.2 | XP_006231458.1 | 8, 9 | 1068 | |||
| XM_008760424.1 | XP_008758646.1 | 8, 9 | 1070 | |||
| XM_008760425.1 | XP_008758647.1 | 8, 9 | 833 | |||
| GenBank mRNA | BC079368.1 | - | 8, 9 | 796 | 7109628 | |

Pyroxd2
Genomic Map