siRNA Details
| Gene Symbol: Cox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101929.1 | - | 8 | 1141 | 7939237 | |
| Gene Symbol: LOC100911779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749128.3 | XP_003749176.2 | 1783 | |||

Cox15
Genomic Map