siRNA Details
| Gene Symbol: Abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014028.1 | NP_001014050.1 | 2 | 523 | ||
| XM_006231162.2 | XP_006231224.1 | 2 | 613 | |||
| GenBank mRNA | BC079229.1 | - | 2 | 523 | 7132456 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146096, AK050879.

Abhd17b
Genomic Map